2-((2-Methoxybenzyl)oxy)acetic acid 95%

Supplier: Ambeed
Warning

A1093269-1G A1093269-100MG A1093269-250MG
76672-170EA 227 USD
76672-170 77783-179 77783-180
2-((2-Methoxybenzyl)oxy)acetic acid 95%
2-((2-Methoxybenzyl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD19370082
CAS Number: 180044-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR