2-Fluoro-5-(N-isopropyl-N-methylsulfamoyl)benzoic acid 95%

Supplier: Ambeed
Warning

A1091990-100MG A1091990-1G A1091990-250MG
76672-162EA 76.39 USD
76672-162 76672-164 76672-166
2-Fluoro-5-(N-isopropyl-N-methylsulfamoyl)benzoic acid 95%
2-Fluoro-5-(N-isopropyl-N-methylsulfamoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO₄S
MW: 275.2966 g/mol
MDL Number: MFCD09041208
CAS Number: 926191-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR