1-Bromo-3-(3-bromopropoxy)propane 95%

Supplier: Ambeed
Warning

A1091550-100MG A1091550-1G A1091550-250MG
76672-156EA 77.45 USD
76672-156 76672-158 76672-160
1-Bromo-3-(3-bromopropoxy)propane 95%
1-Bromo-3-(3-bromopropoxy)propane

Precaución: for Research Use Only

Formula: C₆H₁₂Br₂O
MW: 259.9669 g/mol
MDL Number: MFCD22200244
CAS Number: 58929-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR