3-(Aminomethyl)-2-fluorophenol hydrobromide 95%

Supplier: Ambeed
Warning

A1091445-100MG A1091445-1G A1091445-250MG
76672-150EA 101.85 USD
76672-150 76672-152 76672-154
3-(Aminomethyl)-2-fluorophenol hydrobromide 95%
3-(Aminomethyl)-2-fluorophenol hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉BrFNO
MW: 222.0549 g/mol
MDL Number: MFCD28960657
CAS Number: 1143571-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR