1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1091390-1G
A1091390-250MG
76672-146EA
308.73
USD
76672-146
76672-148
1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride 95%
1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₂H₁₃ClN₂O₂ MW: 252.6968 g/mol |
MDL Number:
MFCD09971425 CAS Number: 1156899-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...