7-Pentadecanol 95%

Supplier: Ambeed
Warning

Synonyms: Pentadecan-7-ol

A1090687-1G A1090687-250MG A1090687-5G
76672-140EA 136.86 USD
76672-140 76672-142 77565-920
7-Pentadecanol 95%
7-Pentadecanol

Precaución: For research use only.

Formula: C₁₅H₃₂O
MW: 228.4140 g/mol
MDL Number: MFCD00046732
CAS Number: 4104-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR