3-(3-Bromophenyl)prop-2-ynoic acid 95%

Supplier: Ambeed
Warning

A1089404-100MG A1089404-1G A1089404-250MG
76672-132EA 25 USD
76672-132 76672-134 76672-136
3-(3-Bromophenyl)prop-2-ynoic acid 95%
3-(3-Bromophenyl)prop-2-ynoic acid

Precaución: for Research Use Only

Formula: C₉H₅BrO₂
MDL Number: MFCD15147227
CAS Number: 29835-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR