2,7-Dichloroimidazo[1,2-a]pyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A1088889-100MG A1088889-1G A1088889-250MG
76672-122EA 81.69 USD
76672-122 76672-124 76672-126
2,7-Dichloroimidazo[1,2-a]pyridine-3-carboxylic acid 95%
2,7-Dichloroimidazo[1,2-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂O₂
MW: 231.0356 g/mol
MDL Number: MFCD29991782
CAS Number: 1955547-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR