(3-Bromo-4-fluorophenyl)methanesulfonamide 95%

Supplier: Ambeed
Warning

A1086053-1G A1086053-250MG
76672-108EA 254.62 USD
76672-108 76672-110
(3-Bromo-4-fluorophenyl)methanesulfonamide 95%
(3-Bromo-4-fluorophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₇BrFNO₂S
MW: 268.1034 g/mol
MDL Number: MFCD12783426
CAS Number: 1184814-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR