2-Chloro-4-fluorobenzenemethanesulfonamide 95%

Supplier: Ambeed
Warning

A1085951-100MG A1085951-1G
76672-104EA 23 USD
76672-104 76672-106
2-Chloro-4-fluorobenzenemethanesulfonamide 95%
2-Chloro-4-fluorobenzenemethanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₇ClFNO₂S
MDL Number: MFCD11212986
CAS Number: 1001026-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR