1-(1-Methyl-1H-pyrazol-3-yl)ethan-1-ol 98%

Supplier: Ambeed
Warning

A1085006-100MG A1085006-1G A1085006-250MG
76672-102EA 26 USD
76672-102 77592-164 77592-166
1-(1-Methyl-1H-pyrazol-3-yl)ethan-1-ol 98%
1-(1-Methyl-1H-pyrazol-3-yl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MDL Number: MFCD11035749
CAS Number: 60031-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR