2-[2-(tert-Butoxy)-2-oxoethoxy]acetic acid 95%

Supplier: Ambeed
Warning

A1083279-1G A1083279-250MG A1083279-5G
76672-074EA 247.19 USD
76672-074 76672-076 76672-078
2-[2-(tert-Butoxy)-2-oxoethoxy]acetic acid 95%
2-[2-(tert-Butoxy)-2-oxoethoxy]acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₅
MW: 190.1938 g/mol
MDL Number: MFCD20640417
CAS Number: 120289-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR