Methyl 2-(2,5-dioxoimidazolidin-4-yl)acetate 95%

Supplier: Ambeed
Warning

A1082980-100MG A1082980-1G
76672-070EA 5 USD
76672-070 76672-072
Methyl 2-(2,5-dioxoimidazolidin-4-yl)acetate 95%
Methyl 2-(2,5-dioxoimidazolidin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₄
MW: 172.14 g/mol
MDL Number: MFCD05884945
CAS Number: 78212-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR