5-Bromo-3-cyclopropyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

A1082338-100MG A1082338-250MG A1082338-50MG A1082338-1G
76672-052EA 118.83 USD
76672-052 76672-054 76672-056 77595-734
5-Bromo-3-cyclopropyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-3-cyclopropyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂
MW: 237.0959 g/mol
MDL Number: MFCD18258924
CAS Number: 1256818-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR