3-Fluoro-4-methoxypyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1082330-100MG A1082330-250MG
76672-048EA 128.37 USD
76672-048 76672-050
3-Fluoro-4-methoxypyridine-2-carboxylic acid 95%
3-Fluoro-4-methoxypyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MW: 171.13 g/mol
MDL Number: MFCD18257407
CAS Number: 1256810-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR