1-[2-(Dimethylamino)ethyl]-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(Dimethylamino)ethyl)-1H-pyrazole-4-carboxylic acid

A1082074-250MG A1082074-1G A1082074-100MG
76672-046EA 320.4 USD
76672-046 77595-732 77619-804
1-[2-(Dimethylamino)ethyl]-1H-pyrazole-4-carboxylic acid 95%
1-[2-(Dimethylamino)ethyl]-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O₂
MW: 183.2077 g/mol
MDL Number: MFCD12139631
CAS Number: 874196-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR