1-[(tert-Butoxy)carbonyl]-2-oxopyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A1080716-100MG A1080716-1G A1080716-250MG
76672-026EA 36 USD
76672-026 76672-028 76672-030
1-[(tert-Butoxy)carbonyl]-2-oxopyrrolidine-3-carboxylic acid 95%
1-[(tert-Butoxy)carbonyl]-2-oxopyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₅
MDL Number: MFCD30209256
CAS Number: 1370018-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR