Methyl 2-(dichloromethyl)-4,5-dihydro-1,3-oxazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A1079845-1G A1079845-250MG A1079845-5G
76672-020EA 63 USD
76672-020 76672-022 76672-024
Methyl 2-(dichloromethyl)-4,5-dihydro-1,3-oxazole-4-carboxylate 95%
Methyl 2-(dichloromethyl)-4,5-dihydro-1,3-oxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇Cl₂NO₃
MDL Number: MFCD29041151
CAS Number: 289030-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR