2-(2-Methylpyridin-4-yl)-1,3-oxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A1079592-100MG A1079592-1G A1079592-250MG A1079592-50MG
76672-008EA 263.11 USD
76672-008 76672-010 76672-012 76672-014
2-(2-Methylpyridin-4-yl)-1,3-oxazole-4-carboxylic acid 98%
2-(2-Methylpyridin-4-yl)-1,3-oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₃
MW: 204.1821 g/mol
MDL Number: MFCD18261012
CAS Number: 1256786-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR