1-(Pyridin-4-yl)pentan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A1078130-100MG A1078130-1G A1078130-250MG
76672-002EA 163.38 USD
76672-002 76672-004 76672-006
1-(Pyridin-4-yl)pentan-1-amine hydrochloride 95%
1-(Pyridin-4-yl)pentan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MW: 200.7084 g/mol
MDL Number: MFCD28126434
CAS Number: 1864057-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR