2-(Furan-2-yl)pyridin-4-amine 95%

Supplier: Ambeed
Warning

A1077284-100MG A1077284-1G A1077284-250MG
76671-994EA 101.85 USD
76671-994 76671-996 76671-998
2-(Furan-2-yl)pyridin-4-amine 95%
2-(Furan-2-yl)pyridin-4-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD12032854
CAS Number: 1159817-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR