1-Piperazin-1-ylacetone dihydrochloride 95%

Supplier: Ambeed
Warning

A1077237-100MG A1077237-1G A1077237-250MG
76671-984EA 60 USD
76671-984 76671-986 76671-988
1-Piperazin-1-ylacetone dihydrochloride 95%
1-Piperazin-1-ylacetone dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂O
MW: 215.1207 g/mol
MDL Number: MFCD21090476
CAS Number: 1353504-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR