Hydrazine, [[4-(phenylmethoxy)phenyl]methyl]-, hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1076968-100MG
A1076968-1G
A1076968-250MG
A1076968-5G
76671-976EA
16
USD
76671-976
76671-978
76671-980
76671-982
Hydrazine, [[4-(phenylmethoxy)phenyl]methyl]-, hydrochloride 95%
Hydrazine, [[4-(phenylmethoxy)phenyl]methyl]-, hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₇ClN₂O |
MDL Number:
MFCD12031972 CAS Number: 237064-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...