4-(Morpholine-4-sulfonyl)-benzoyl chloride 95%

Supplier: Ambeed
Danger

A1076677-100MG A1076677-1G A1076677-250MG
76671-970EA 72.15 USD
76671-970 76671-972 76671-974
4-(Morpholine-4-sulfonyl)-benzoyl chloride 95%
4-(Morpholine-4-sulfonyl)-benzoyl chloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₄S
MW: 289.7353 g/mol
MDL Number: MFCD09702532
CAS Number: 654682-86-7
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR