Methyl 4-(1-benzofuran-2-yl)-2,4-dioxobutanoate 95%

Supplier: Ambeed
Warning

A1076510-100MG A1076510-1G A1076510-250MG
76671-964EA 80.63 USD
76671-964 76671-966 76671-968
Methyl 4-(1-benzofuran-2-yl)-2,4-dioxobutanoate 95%
Methyl 4-(1-benzofuran-2-yl)-2,4-dioxobutanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₅
MW: 246.2155 g/mol
MDL Number: MFCD06496404
CAS Number: 889996-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR