Benzyl (2S,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A1074723-100MG A1074723-250MG
76671-946EA 78.51 USD
76671-946 77619-798
Benzyl (2S,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 95%
Benzyl (2S,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO₃
MW: 257.7133 g/mol
MDL Number: MFCD29034383
CAS Number: 1864003-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR