N-(4-[(2E)-3-(dimethylamino)prop-2-enoyl]phenyl)acetamide 95%

Supplier: Ambeed
Warning

A1074032-1G A1074032-250MG
76671-942EA 123 USD
76671-942 76671-944
N-(4-[(2E)-3-(dimethylamino)prop-2-enoyl]phenyl)acetamide 95%
N-(4-[(2E)-3-(dimethylamino)prop-2-enoyl]phenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MDL Number: MFCD08559039
CAS Number: 1056165-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR