1-Iodo-2-methyl-4-(trifluoromethyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-3-methylbenzotrifluoride

A1073354-100MG A1073354-1G A1073354-250MG A1073354-5G
76671-932EA 67.9 USD
76671-932 76671-934 76671-936 76671-938
1-Iodo-2-methyl-4-(trifluoromethyl)benzene 95%
1-Iodo-2-methyl-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆F₃I
MW: 286.0329 g/mol
MDL Number: MFCD22570417
CAS Number: 54978-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR