N-Benzyl-2-acetamidoprop-2-enamide 95%

Supplier: Ambeed
Warning

A1072859-100MG A1072859-250MG A1072859-5G
76671-918EA 91.24 USD
76671-918 76671-920 76671-922
N-Benzyl-2-acetamidoprop-2-enamide 95%
N-Benzyl-2-acetamidoprop-2-enamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.2518 g/mol
MDL Number: MFCD24550141
CAS Number: 86921-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR