Methyl 3-(fluorosulfonyl)benzoate 95%

Supplier: Ambeed
Danger

A1072562-100MG A1072562-1G A1072562-250MG
76671-910EA 13 USD
76671-910 76671-912 76671-914
Methyl 3-(fluorosulfonyl)benzoate 95%
Methyl 3-(fluorosulfonyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₄S
MW: 218.2022 g/mol
MDL Number: MFCD22192516
CAS Number: 124397-36-0
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR