Decahydronaphthalene-1,8-dione 95%

Supplier: Ambeed
Warning

A1072166-1G A1072166-250MG A1072166-100MG
76671-906EA 390 USD
76671-906 77570-430 77783-096
Decahydronaphthalene-1,8-dione 95%
Decahydronaphthalene-1,8-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MDL Number: MFCD24689133
CAS Number: 83406-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR