2-Amino-4-chloro-3-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A1072031-100MG A1072031-1G A1072031-250MG
76671-900EA 128.37 USD
76671-900 76671-902 76671-904
2-Amino-4-chloro-3-hydroxybenzoic acid 95%
2-Amino-4-chloro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.5804 g/mol
MDL Number: MFCD20639114
CAS Number: 23219-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR