2-(3-Amino-4-methoxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A1071150-1G A1071150-250MG
76671-882EA 407.39 USD
76671-882 76671-884
2-(3-Amino-4-methoxyphenyl)acetic acid 95%
2-(3-Amino-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.1885 g/mol
MDL Number: MFCD12913543
CAS Number: 63304-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR