2-Bromo-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one 95%

Supplier: Ambeed
Danger

A1070975-1G A1070975-250MG A1070975-5G
76671-880EA 46 USD
76671-880 77783-090 77783-091
2-Bromo-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one 95%
2-Bromo-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MDL Number: MFCD15206027
CAS Number: 145797-86-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR