3-(4-Methyl-6-oxo-1,6-dihydropyrimidin-1-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A1070906-100MG A1070906-1G A1070906-250MG
76671-874EA 173.99 USD
76671-874 76671-876 76671-878
3-(4-Methyl-6-oxo-1,6-dihydropyrimidin-1-yl)propanoic acid 95%
3-(4-Methyl-6-oxo-1,6-dihydropyrimidin-1-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.1766 g/mol
MDL Number: MFCD14601704
CAS Number: 1249567-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR