5-Iodopent-1-ene 95%

Supplier: Ambeed
Danger

A1069531-1G A1069531-250MG A1069531-5G
76671-838EA 68.96 USD
76671-838 76671-840 77592-144
5-Iodopent-1-ene 95%
5-Iodopent-1-ene

Precaución: for Research Use Only

Formula: C₅H₉I
MW: 196.03 g/mol
MDL Number: MFCD19688471
CAS Number: 7766-48-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR