tert-Butyl (1-(hydroxymethyl)cyclopropyl)(methyl)carbamate 95%

Supplier: Ambeed
Warning

A1069451-1G A1069451-100MG A1069451-250MG
76671-836EA 595 USD
76671-836 77783-085 77783-086
tert-Butyl (1-(hydroxymethyl)cyclopropyl)(methyl)carbamate 95%
tert-Butyl (1-(hydroxymethyl)cyclopropyl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD24467438
CAS Number: 387845-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR