Methyl 6-fluoro-2-oxo-2,3-dihydro-1H-indole-5-carboxylate 95%

Supplier: Ambeed
Danger

A1069182-1G A1069182-250MG
76671-832EA 1059.84 USD
76671-832 76671-834
Methyl 6-fluoro-2-oxo-2,3-dihydro-1H-indole-5-carboxylate 95%
Methyl 6-fluoro-2-oxo-2,3-dihydro-1H-indole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₃
MW: 209.1700 g/mol
MDL Number: MFCD24617111
CAS Number: 1265965-97-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR