Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A1068557-1G A1068557-250MG
76671-808EA 908.14 USD
76671-808 76671-810
Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate 95%
Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrN₃O₂
MW: 270.0827 g/mol
MDL Number: MFCD23708550
CAS Number: 1427376-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR