2-(4-Bromophenyl)oxetane 95%

Supplier: Ambeed
Warning

A1068550-100MG A1068550-250MG A1068550-50MG
76671-800EA 77 USD
76671-800 76671-802 76671-804
2-(4-Bromophenyl)oxetane 95%
2-(4-Bromophenyl)oxetane

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD31420690
CAS Number: 25574-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR