2-(3-Cyano-5-fluorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A1068509-100MG A1068509-1G
76671-796EA 129.43 USD
76671-796 76671-798
2-(3-Cyano-5-fluorophenyl)acetic acid 95%
2-(3-Cyano-5-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.1479 g/mol
MDL Number: MFCD18394074
CAS Number: 1261823-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR