2-Fluoro-5-methylbenzeneboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1,3,2-Dioxaborolane, 2-(2-fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-, 2-(2-Fluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-methylphenylboronic acid pinacol ester

A1067863-100MG A1067863-1G A1067863-250MG A1067863-5G
76671-786EA 36.08 USD
76671-786 76671-788 76671-790 76671-792
2-Fluoro-5-methylbenzeneboronic acid pinacol ester 95%
2-Fluoro-5-methylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
MDL Number: MFCD12923174
CAS Number: 1192045-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR