(1-((Dimethylamino)methyl)cyclopropyl)methanol 95%

Supplier: Ambeed
Danger

A1066535-100MG A1066535-1G A1066535-250MG A1066535-50MG
76671-766EA 24 USD
76671-766 76671-768 76671-770 76671-772
(1-((Dimethylamino)methyl)cyclopropyl)methanol 95%
(1-((Dimethylamino)methyl)cyclopropyl)methanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MDL Number: MFCD19313167
CAS Number: 39943-41-4
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR