2-(1H-Imidazol-4-yl)pyridine hydrochloride 95%

Supplier: Ambeed
Warning

A1066378-100MG A1066378-1G A1066378-250MG A1066378-5G A1066378-10G
76671-758EA 37 USD
76671-758 76671-760 76671-762 76671-764 77783-066
2-(1H-Imidazol-4-yl)pyridine hydrochloride 95%
2-(1H-Imidazol-4-yl)pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₈ClN₃
MDL Number: MFCD22056293
CAS Number: 1955493-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR