5-Aminopentane-1-thiol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5-aminopentane-1-thiolhydrochloride

A1065972-100MG A1065972-250MG
76671-752EA 142.17 USD
76671-752 76671-754
5-Aminopentane-1-thiol hydrochloride 95%
5-Aminopentane-1-thiol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄ClNS
MW: 155.6894 g/mol
MDL Number: MFCD28383723
CAS Number: 31110-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR