Sodium 2,3-dihydro-1,4-benzodioxine-6-sulfinate 95%

Supplier: Ambeed
Warning

A1065724-100MG A1065724-1G A1065724-250MG
76671-746EA 72 USD
76671-746 76671-748 76671-750
Sodium 2,3-dihydro-1,4-benzodioxine-6-sulfinate 95%
Sodium 2,3-dihydro-1,4-benzodioxine-6-sulfinate

Precaución: for Research Use Only

Formula: C₈H₇NaO₄S
MDL Number: MFCD24113278
CAS Number: 1514973-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR