Sodium 4-nitrobenzene-1-sulfinate 95%

Supplier: Ambeed
Warning

A1065264-1G A1065264-250MG
76671-744EA 119 USD
76671-744 77783-062
Sodium 4-nitrobenzene-1-sulfinate 95%
Sodium 4-nitrobenzene-1-sulfinate

Precaución: for Research Use Only

Formula: C₆H₄NNaO₄S
MDL Number: MFCD24113272
CAS Number: 15959-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR