Hexahydro-1H-pyrrolizin-7a-ylmethanol 98%

Supplier: Ambeed
Warning

A1064282-1G A1064282-250MG A1064282-25G A1064282-5G
76671-716EA 31.83 USD
76671-716 76671-718 76671-720 76671-722
Hexahydro-1H-pyrrolizin-7a-ylmethanol 98%
Hexahydro-1H-pyrrolizin-7a-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₁₅NO
MW: 141.2108 g/mol
MDL Number: MFCD11878488
CAS Number: 78449-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR