Oxepane-4-carboxylic acid 98%

Supplier: Ambeed
Danger

A1064243-100MG A1064243-1G A1064243-250MG
76671-714EA 129 USD
76671-714 77570-380 77570-382
Oxepane-4-carboxylic acid 98%
Oxepane-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MDL Number: MFCD21124931
CAS Number: 933747-23-0
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR