1-(4-Chlorophenyl)-5-methylimidazolidine-2,4-dione 95%

Supplier: Ambeed
Warning

A1063986-1G A1063986-100MG A1063986-250MG
76671-710EA 127 USD
76671-710 77783-044 77783-045
1-(4-Chlorophenyl)-5-methylimidazolidine-2,4-dione 95%
1-(4-Chlorophenyl)-5-methylimidazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD06680124
CAS Number: 1008068-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR